Anti-IgG2a Goat Polyclonal Antibody (CF™ 770)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20264 20264-1
20264.EA 3130 DKK
20264. 20264-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 770)
Antistoffer
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse