Anti-IgG Goat Polyclonal Antibody (Biotin)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20186 20186-1
20186.EA 2680 DKK
20186. 20186-1
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR