Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20117 20117-1
20117.EA 2240 DKK
20117. 20117-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Antistoffer
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat