Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20097 20097-1
20097.EA 2680 DKK
20097. 20097-1
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human