Anti-IgG Donkey Polyclonal Antibody (CF™ 568)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20092 20092-1
20092.EA 2680 DKK
20092. 20092-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR