Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20091 20091-1
20091.EA 2240 DKK
20091. 20091-1
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antistoffer
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR