Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20063 20063-1
20063.EA 2160 DKK
20063. 20063-1
Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Antistoffer
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™680 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red/near-infrared CF™680 (Ex/Em 681/698 nm) is brighter and provides the best signal-to-noise ratio compared to antibody conjugates of spectrally similar dyes.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF680
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse