Anti-IgG Goat Polyclonal Antibody (CF™ 660C)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20053 20053-1
20053.EA 2240 DKK
20053. 20053-1
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660C Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR