Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20039 20039-1
20039.EA 2680 DKK
20039. 20039-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antistoffer
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat