Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20015 20015-1
20015.EA 2680 DKK
20015. 20015-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit