Anti-PICK1 Rabbit Monoclonal Antibody [clone: EPR4130(3)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB133773-100 AB133773-1ML
ABCAAB133773-100EA 4850 DKK
ABCAAB133773-100 ABCAAB133773-1ML
Anti-PICK1 Rabbit Monoclonal Antibody [clone: EPR4130(3)]
Antistoffer
Advarsel:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: PICK1 (protein interacting with PRKCA 1)
Clonality: monoclonal
Clone: EPR4130
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR