Anti-IL-12A Rabbit Monoclonal Antibody [clone: EPR5736]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB133751-100 AB133751-10 AB133751-1ML
ABCAAB133751-100EA 5090 DKK
ABCAAB133751-100 ABCAAB133751-10 ABCAAB133751-1ML
Anti-IL-12A Rabbit Monoclonal Antibody [clone: EPR5736]
Antistoffer
Rabbit monoclonal [EPR5736] to IL-12A.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: IL-12A
Clonality: Monoclonal
Clone: EPR5736
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human