Anti-Cleaved PARP1 Rabbit Monoclonal Antibody [clone: Y34]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB32561-100 AB32561-1ML AB32561-20UL
ABCAAB32561-100EA 3550 DKK
ABCAAB32561-100 ABCAAB32561-1ML ABCAAB32561-20UL
Anti-Cleaved PARP1 Rabbit Monoclonal Antibody [clone: Y34]
Antistoffer
Rabbit monoclonal [Y34] to Cleaved PARP1.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cleaved PARP1
Clonality: Monoclonal
Clone: Y34
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR