Anti-VEGF Receptor 1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB2350-500 AB2350-100 AB2350-1
ABCAAB2350-500EA 5510 DKK
ABCAAB2350-500 ABCAAB2350-100 ABCAAB2350-1
Anti-VEGF Receptor 1 Rabbit Polyclonal Antibody
Antistoffer
Rabbit polyclonal to VEGF Receptor 1.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: VEGF Receptor 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human