Anti-TRB Armenian Hamster Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: H57-597]
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
65-5961-U100 65-5961-U025
65-5961-U100EA 1320 DKK
65-5961-U100 65-5961-U025
Anti-TRB Armenian Hamster Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: H57-597]
Antistoffer
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death.
The H57-597 antibody is used as a phenotypic marker for T cells expressing the alpha-beta TCR. It is also widely used to cross-link surface TCR and thereby mimic TCR-mediated cell activation or induction of apoptosis. The antibody does not cross-react with cells expressing the g/d TCR.
Type: Primary
Antigen: TRB (T cell receptor beta locus)
Clonality: monoclonal
Clone: H57-597
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse