Anti-CD3D Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UCHT1]

Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

60-0038-T100 60-0038-T025
60-0038-T100EA 1180 DKK
60-0038-T100 60-0038-T025
Anti-CD3D Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UCHT1]
Antistoffer
The UCHT1 antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing cell proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.

The UCHT1 antibody is a widely used phenotypic marker for human T cells. In addition, binding/cross-linking of UCHT1 antibody to CD3e can induce cell activation (use format suitable for functional assays). A recent publication of the crystal structure of a CD3e- antibody complex provides insight as to the action of commonly used agonist antibodies, as well as specific epitope-binding data for the human CD3 antibodies UCHT1 and OKT3 (Fernandes, R.A. et al. 2012. J. Biol. Chem. 287: 13324-13335).

Type: Primary
Antigen: CD3D (CD3d molecule, delta (CD3-TCR complex))
Clonality: monoclonal
Clone: UCHT1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR