Sodium hydroxide solution 0,5 mol/L 0.5 mol/l, EMPROVE® EXPERT for biopharmaceutical production
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Caustic soda
1.37060.1000 1.37060.9950
1.37060.1000EA 740 DKK
1.37060.1000 1.37060.9950
Sodium hydroxide solution 0,5 mol/L 0.5 mol/l, EMPROVE® EXPERT for biopharmaceutical production
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: ∼105 °C (1013 hPa)
Smeltepunkt: 323
Massefylde: 1.02 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8
Merck Index: 14,08627