Natriumchlorid solution 3 mol/l, EMPROVE® EXPERT for biopharmaceutical production
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Saline, NaCl, Sea salt, Halite
1.37076.1000
1.37076.9010
1.37076.1000EA
847
DKK
1.37076.1000
1.37076.9010
Natriumchlorid solution 3 mol/l, EMPROVE® EXPERT for biopharmaceutical production
Sodium chloride
Formel:
NaCl Kogepunkt: 109 °C (1013 hPa) Massefylde: ≤1,11 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003477 CAS-nummer: 7647-14-5 EINECS: 215-185-5 UN nummer: 1824 ADR: 8,II |