ortho-Phosphorsyre 1 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
4.80939.1000
4.80939.9950
4.80939.1000EA
497
DKK
4.80939.1000
4.80939.9950
ortho-Phosphorsyre 1 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
ortho-Phosphorsyre
Formel:
H₃PO₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011340 CAS-nummer: 7664-38-2 EINECS: 231-633-2 UN nummer: 1805 ADR: 8,III |