Safety spectacles, SecureFit™ 400
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SF408AS SF401AF SF402AF SF403AF SF410AS SF420AF-EU SF425AF-EU SF415AF-EU
111-1614EA 138 DKK
111-1614 111-1611 111-1612 111-1613 111-1615 MMMASF420AF-EU MMMASF425AF-EU MMMASF415AF-EU
Safety spectacles, SecureFit™ 400
Briller
Sinlge lens safety glasses in a modern design that provides a combination of high comfort and an optimal fit without any feeling of pressure.
  • Temple pressure distribution ensures a soft feel
  • Dual-injected, padded temple touchpoints for added comfort over the ears
  • Special bracket design for natural adaptation to different head sizes
  • Soft, adjustable nose pads for individual fit
  • Polycarbonate lenses that absorb 99,9% of UVA and UVB rays
Weight: 19 g