Industrial gloves, oil repellent, HyFlex® 11-926
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-926/11 11-926/8 11-926/6 11-926/7 11-926/9 11-926/10
112-4291EA 887 DKK
112-4291 112-4294 112-4292 112-4293 112-4295 112-4290
Industrial gloves, oil repellent, HyFlex® 11-926
Handsker
This knitted Spandex/nylon glove features a ¾ dipped neoprene/nitrile coating which provides oil repellence. Ideal for use in oily environments.
  • RIPEL™ technology creates a liquid-impermeable layer that prevents oil or lubricant from making even incidental contact with the wearer’s skin
  • Durable ¾ dipped coating ensures full protection for the back of the fingers and a safe grip in oily environments
  • Liner is made with one of the finest gauge nylon fibre
  • Enhanced flexibility and tactility
Suitable applications include: Stamping light and small metal parts, primary assembly of light components, maintenance operations, loading and downloading parts in metal working lines and assembly in machining operations.
Certificeringer: EN 388:2016 (4121A); Cat. II.