HPLC columns, Hibar®, with Aluspher® sorbents
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00424.7002 1.50182.7002 1.50183.7002
1.00424.7002EA 0 DKK
1.00424.7002 1.50182.7002 1.50183.7002
HPLC columns, Hibar®, with Aluspher® sorbents
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with Aluspher® sorbents. Ideal for separation of compounds under basic eluents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.