HPLC columns, Hibar®, with LiChrosorb® sorbents
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00423.7054 1.00423.7057 1.00424.7054 1.00424.7057 1.00424.7059 1.50009.7052 1.50009.7054 1.50009.7057 1.50009.7059 1.50012.7052 1.50012.7054 1.50012.7059 1.50013.7052 1.50013.7054 1.50013.7057 1.50183.7052 1.50183.7054 1.50183.7057 1.50183.7059 1.50013.7059
1.00423.7054EA 7340 DKK
1.00423.7054 1.00423.7057 1.00424.7054 1.00424.7057 1.00424.7059 1.50009.7052 1.50009.7054 1.50009.7057 1.50009.7059 1.50012.7052 1.50012.7054 1.50012.7059 1.50013.7052 1.50013.7054 1.50013.7057 1.50183.7052 1.50183.7054 1.50183.7057 1.50183.7059 1.50013.7059
HPLC columns, Hibar®, with LiChrosorb® sorbents
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with LiChrosorb® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.