Test tube racks, Poxygrid™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F187720001 F187590001 F187500001 F187530001 F187640001
119-0456EA 904 DKK
119-0456 119-0453 119-0450 119-0451 119-0455
Test tube racks, Poxygrid™, SP Bel-Art
Stativer Tube Racks
Epoxy coated wire
  • Resistant to organic solvents, salts, weak acids and weak alkalis
  • Fully functional between temperatures of −223 °C to autoclaving at 121 °C
  • Autoclavable
All racks have square holes, three decks, a slotted grid bottom and reinforced edges.