Industrial gloves, RewoMech® 640
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
640.10 640.11 640.12 640.7 640.8 640.9
KCHL640.10EA 249 DKK
KCHL640.10 KCHL640.11 KCHL640.12 KCHL640.7 KCHL640.8 KCHL640.9
Industrial gloves, RewoMech® 640
Handsker
These extremely resistant artificial leather gloves offer good mechanical protection. The glove's back is made of light elastic textile for a perfect fit.
  • Good dexterity
  • No material hardening
  • Absorbent surface profile for a good grip
  • Back of the thumb is equipped with the TerryCotton fabric for sweat absorption
  • Cuff with Velcro® fastener for perfect fit
  • Washable
Ideal for metal and plastics processing, mechanical engineering, automotive and supply industry cleaning and maintenance, logistics and when working with oily and wet parts.
Certificeringer: EN 388:2016 (2121X); Cat. II.