Industrial gloves, Camapur® Comfort 619
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
619.10 619.9 619.8 619.6 619.7
KCHL619.10EA 71.5 DKK
KCHL619.10 KCHL619.9 KCHL619.8 KCHL619.6 KCHL619.7
Industrial gloves, Camapur® Comfort 619
Handsker
These seamless polyamide liners feature a PU palm coating which provides excellent grip when handling greasy and oily parts.
  • Excellent fit
  • Very good sensitivity, very breathable
  • Good mechanical resistance
  • Grey colour for longer service life
  • Washable
Ideal for use in the electronics and automotive industry, for precision mechanics, metal and plastics processing and for storage and commission work.
Certificeringer: EN 388:2016 (3131X); Cat. II