Chemical resistant gloves, AlphaTec® 37-501
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37-501/6.5 37-501/7.5 37-501/8.5 37-501/9.5 37-501/10.5
ANSE37-501/6.5EA 546 DKK
ANSE37-501/6.5 ANSE37-501/7.5 ANSE37-501/8.5 ANSE37-501/9.5 ANSE37-501/10.5
Chemical resistant gloves, AlphaTec® 37-501
Handsker
These nitrile gloves with cotton flock lining provide excellent resistance to many solvents and oil based chemicals.
  • Very good abrasion resistance
  • Flexible and comfortable for an excellent dexterity
  • Open cuff allows ventilation to keep the hand cool
Ideal for use when handling agrochemicals, meat, fruits and vegetables. Also suitable for chemical processing and preparation, degreasing, maintenance, metal fabrication, tank filling and packaging applications.
Certificeringer: EN ISO 374-1:2016 Type A (AJKLPT), EN ISO 374-5:2016, EN 388:2016 (3101X); Cat. III; AQL 0,65
Food contact approved