Nitrile gloves, TouchNTuff® 93-250
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
93-250/8.5-9 93-250/9.5-10 93-250/5.5-6 93-250/6.5-7 93-250/7.5-8
112-4232EA 574 DKK
112-4232 112-4233 112-4776 112-4230 112-4231
Nitrile gloves, TouchNTuff® 93-250
Handsker
These nitrile gloves resist a variety of industrial chemicals for longer periods than other nitrile disposable gloves. They also offer high tensile strength and 3 times the puncture resistance of latex or PVC gloves.
  • Non sterile, powder-free, textured fingertips
  • Ansell Grip Technology™ minimises the force required to grip dry, wet or oily objects, reducing hand and arm fatigue
  • Proprietary nitrile formulation provides comfort for long wear periods
  • Contains no natural rubber proteins, therefore protects against Type I skin allergies
  • No transfer of silicone