Culture media in bags, Dilubags®
Leverandør: bioMérieux
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AEB911495/2 AEB911493/4
AEB911495/2EA 582 DKK
AEB911495/2 AEB911493/4
Culture media in bags, Dilubags®
Mikrobiologiske medier Klargjort medium
To optimise the sample preparation, the majority of diluents are available in 3 or 5 litre Dilubags® (dilution broth bags).
  • For food industry