Conductivity sensors, InLab® Trace
Leverandør: Mettler - Toledo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30014097 30014099
663-0264EA 17260 DKK
663-0264 663-0265
Conductivity sensors, InLab® Trace
Elektroder Elektroder til ledningsevnemåling
This digital conductivity sensor has a high precision temperature probe. The conductivity and temperature signals are digitised and combined through a special algorithm for unmatched conductivity accuracy.
  • Optional PP flow cell provides protection from carbon dioxide: Ensures very high repeatability and reproducibility
  • Flow cell was built at an angle to prevent air bubbles being trapped
  • Special temperature compensation mode to increase the reliability of readings
  • Certified cell constant makes calibration superfluous
Ideal for low conductivity measurements in pure or ultra-pure water. Suitable for use with SevenExcellence™ and SevenCompact™ meters.
Leveringsinformation: Each sensor is supplied with a certificate specifying the actual cell constant. The electrode is supplied without flow cell. The kit includes the flow cell.