Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4990P-500 4990P-250 4990P-1L 4990P-100
CORN4990P-500EA 863 DKK
CORN4990P-500 CORN4990P-250 CORN4990P-1L CORN4990P-100
Erlenmeyer flasks, narrow neck, with screw cap, Corning®
Kolber Erlenmeyer-kolber
PMP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With easy to read red markings.