PCR microtube strips, 0,1 ml
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030124.804 0030124.812 0030124.820
732-1615EA 1150 DKK
732-1615 732-1616 732-1617
PCR microtube strips, 0,1 ml
Rør PCR rør
Eight reaction tubes in strip-format, ideal for small sample volumes.
  • Easily adaptable for automation
  • Sealable using flat or domed strip-lids
  • Certified free of human DNA, DNase, RNase and PCR Inhibitors