Knitted gloves, HyFlex® 11-800
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-800/7 11-800/8 11-800/9 11-800/10 11-800/6 11-800/11
112-0207EA 611 DKK
112-0207 112-0208 112-0209 112-0205 112-0206 112-4253
Knitted gloves, HyFlex® 11-800
Handsker
This knitted nylon glove features a foam nitrile coating on the palm.
  • Perfect balance between comfort, dexterity and protection
  • Advanced Knitting Technology allows increased stretch and flexibility in high-stress areas so the hand operates more freely, improving comfort and reducing hand fatigue
  • Anti-static according to EN 1149
  • Oeko-Tex® certified
Ideal for precision assembly and general handling in warehouses.
Certificeringer: EN 388:2016 (3131A), EN 1149; Cat. II