Digital block heaters
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88870004 88870005 88870006
460-0306EA 3660 DKK
460-0306 460-0307 460-0308
Digital block heaters
Bloktermostater Digitale bloktermostater
Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducible results. The units have digital controls and display of time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables user to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Interchangeable aluminum alloy sample blocks provide versatility and allow for easy cleaning and disinfecting
Leveringsinformation: Supplied with block extraction tool, but without blocks, these must be ordered separately.
Advarsel: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.