Support for chairs, Allround
Leverandør: Salli Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALLROUND-99999 ALLROUND-11049 ALLROUND-05011
139-0481EA 0 DKK
139-0481 139-0475 139-0474
Support for chairs, Allround
Stole
This support is ideal for precision work in which the working positions change and versatile support is needed for the upper body.
  • Support can be changed from left-handed to right-handed and vice versa
  • Support is attached to the gas cylinder of the chair, so it can be used with all Salli chair models (SwingFit, Swing, MultiAdjuster, Twin, Classic)
  • Support arch length approx. 61 cm, width approx. 8,5 cm
  • Support upholstered with leather or artifical leather
Excellent for dentists, dental surgeons or dental assistants. It can also be used as a stretchingMore Product Information