Microbiology test kit, MEL/MF portable laboratory
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2569700 2580100
702-0011EA 55000 DKK
702-0011 HACH2580100
Microbiology test kit, MEL/MF portable laboratory
Mikrobiologiske testsystemer Test for mikroorganismer
The Total Coliform MEL Laboratory is designed for analysts who need portable or small-scale microbiology testing capabilities. From a remote field location to a small scale microbiology lab within your existing lab, this MEL offers all the equipment and accessories to perform microbiological analysis.
Bestillingsoplysninger: Required media should be ordered separately.
Leveringsinformation: Kit includes portable incubator and rack, thermometer, Whirlpak bags with thiosulfate (200), filter funnel/pad assemblies (200), alcohol burner (order alcohol separately), vacuum filtration apparatus, colony counter, magnifier and permanent marker.