Nitrogen-nitrate standard solution
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2415132 1426010 194749 204649
HACH2415132EA 169 DKK
HACH2415132 705-2010 705-2025 HACH204649
Nitrogen-nitrate standard solution
Standarder Miljø analyse standarder
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.