Spatulas, LaboPlast / SteriPlast
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5378-1009 5378-1010 5378-0009 5378-0010
231-0523EA 243 DKK
231-0523 231-0524 231-0511 231-0512
Spatulas, LaboPlast / SteriPlast
Spatler
PS, white. These disposable spatulas are ideal for the sampling of powder, granules and pastes.
  • Long, ergonomic and solid handle
  • Pointed blade can be easily pierced into paper and plastic bags
  • Available non sterile (LaboPlast) or gamma radiation sterilised (SteriPlast)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
As an option, the spatulas are available with cover in order to secure a contamination-free sampling process.