Sampling spoons, LaboPlast
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5378-0011 5378-0012
231-0513EA 801 DKK
231-0513 231-0514
Sampling spoons, LaboPlast
Skeer
PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.
  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed