Reagent reservoirs
Leverandør: DIVERSIFIED BIOTECH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RESE-1000 RESE-3000 RESE-2000
DIVERESE-1000EA 4280 DKK
DIVERESE-1000 DIVERESE-3000 DIVERESE-2000
Reagent reservoirs
Reagens reservoirs
Disposable reagent reservooirs made of resistant and durable PET.
  • Pour-off spouts on all four corners
  • Wide base and reinforced sidewalls avoid spills and provide stability
  • Sterilised by gamma irradiation
  • 25 ml reservoir with divider accommodates eight channel pipettes on one side and one to four channel pipettes on the other; divider is also convenient for adding controls
  • Optional coolers are available which keep the reagents at ~5 °C for approximately 3 hours