Precision Clean® II ultra clear cleanroom tubing
Leverandør: FISHER CONTAINER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1600080 1600480 1600040
FISH1600080EA 3210 DKK
FISH1600080 FISH1600480 FISH1600040
Precision Clean® II ultra clear cleanroom tubing
Slanger Overførselsslange
This clear Polyethylene tubing features an inner surface cleanliness level suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions.
Bestillingsoplysninger: Individually wrapped and shipped with special plastic cores to prevent particulate contamination.