Seats, Anti-Static, PU
Leverandør: Kango
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7EG20GHLE00905 7EG35GBLN00905 7EG40GHLE01905 7EG35GHLE01905
BIEN7EG20GHLE00905EA 2560 DKK
BIEN7EG20GHLE00905 BIEN7EG35GBLN00905 BIEN7EG40GHLE01905 BIEN7EG35GHLE01905
Seats, Anti-Static, PU
Stole
Flexible PU resistance to sharp tools, sparks, weak acids and hydrocarbons. Special conductive PU connects to the metal parts of the chair, allowing electricity to be discharged through carbon casters or pads.
  • Comfortable and airy surface
  • Without staples/seams
  • Abrasion-resistant
  • Easily adjustable and washable
  • 5-branch scuff resistant reinforced base