Tubing, chloroprene
Leverandør: Saint Gobain Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720390 720480
VERN720390EA 45.5 DKK
VERN720390 VERN720480
Tubing, chloroprene
Slanger Overførselsslange
Chloroprene, black.
  • Resistance to hydrocarbons and mineral oils
  • Good ageing: ozone resistant
  • High di-electrical strength
With its good mechanical, chemical, electrical and heat resistance (thermal) properties, chloroprene is recommended for a wide range of industrial applications. It is used as much for cable insulation, as for transfer of nitrogen and helium gases. It shows an outstanding resistance to UV and weathering.
Temperature resistance: −20 to +100 °C
Hardness (Shore durometer A): 50
Density: 1,55