Slip-on baffle, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C490-12 C490-10
BOHLC490-12EA 237 DKK
BOHLC490-12 BOHLC490-10
Slip-on baffle, BOLA
Omrørere Omrøreudstyr
PTFE, with PFA support ring. The baffle can be mounted at any position on a temperature probe or a solo stirrer shaft.
  • Vacuum suitable
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C
Certificeringer: DIN 28131