Insoles for occupational shoes, 3567 to 3570, Reflexor
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3567/36 3567/37 3567/38 3567/39 3567/40 3567/41 3567/42 3568/36 3568/37 3568/38 3568/39 3568/40 3568/41 3568/42 3569/40 3569/41 3569/42 3569/43 3569/44 3569/45 3569/46 3570/40 3570/41 3570/42 3570/43 3570/44 3570/45 3570/46
ABEB3567/36EA 118 DKK
ABEB3567/36 ABEB3567/37 ABEB3567/38 ABEB3567/39 ABEB3567/40 ABEB3567/41 ABEB3567/42 ABEB3568/36 ABEB3568/37 ABEB3568/38 ABEB3568/39 ABEB3568/40 ABEB3568/41 ABEB3568/42 ABEB3569/40 ABEB3569/41 ABEB3569/42 ABEB3569/43 ABEB3569/44 ABEB3569/45 ABEB3569/46 ABEB3570/40 ABEB3570/41 ABEB3570/42 ABEB3570/43 ABEB3570/44 ABEB3570/45 ABEB3570/46
Insoles for occupational shoes, 3567 to 3570, Reflexor
Sko indlægssåler
Smooth anti-static insoles made from microfibre.
  • Reflexor massage active-integrated insole
  • Comfortable orthopaedic integrated insole
Colour: White or anthracite
Colour: White/anthracite
Certificeringer: EN 61340 (ESD)