Industrial gloves, AlphaTec® 23-202
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23-202/10 23-202/8 23-202/9
ANSE23-202/10EA 780 DKK
ANSE23-202/10 ANSE23-202/8 ANSE23-202/9
Industrial gloves, AlphaTec® 23-202
Handsker
These PVC coated gloves with inner polar acrylic seamless lining provide chemical and liquid protection and good dexterity at low temperatures.
  • Roughened sandblast finish on the glove and smooth finish on the gauntlet cuff
  • Designed to protect at temperatures down to −30 °C
  • Immediate heat feeling when wearing the glove
  • Good grip in handling both slippery and wet materials
  • Soft integral PVC coating offers chemical resistance
Ideal for outdoor applications including chemical handlings.
Gauge: 13
Colour: Blue
EN 388 (4121), EN 374, EN 421, EN 511More Product Information