Chemical resistant gloves, AlphaTec® 23-201
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23-201/10 23-201/7 23-201/8 23-201/9
112-4245EA 1420 DKK
112-4245 112-4244 112-4243 112-4242
Chemical resistant gloves, AlphaTec® 23-201
Handsker
These cotton liners with soft PVC coating and heat-sealed sleeve maintain their dexterity and pliability at low temperatures. They provide chemical, mechanical and liquid protection.
  • Dipped rough finish makes handling wet and slippery materials easier and safer
  • Heat-sealed sleeve offers liquid resistance and dust protection only
Ideal for janitorial use, chemical handling, cleaning applications as well as for fish, fruit and vegetable processing.
Thickness: 1,60 mm
Colour: Blue
EN 374, EN 388 (4121); Cat. III; AQL 1,5 Food contact approved
Certificeringer: EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:2016, EN 388:2016 (3121B); Cat. III; AQL 1,5
Food contact approved.