VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0629EA 584 DKK
525-0629 525-0630 525-0631 525-0632
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Rør Centrifugerør
PP, high grade gamma resistance, conical, with HDPE flat cap.
  • Easy-to-read black graduations are accurate to ±2%, with 0,5 ml increments (15 ml) or 2,5 ml increments (50 ml)
  • With a large unerasable frosted white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Autoclavable at +121 °C and freezable to −80 °C
  • Leakproof
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Ideal for environmental uses such as water analysis and other applications where tubes have the potential to contaminate samples. Tubes are RNase-free, DNase-free, and non pyrogenic.
Certificeringer: TSE/BSE-free Certification.
Leveringsinformation: Racked tubes are packaged in environmentally responsible (fully recyclable) chipboard racks. Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability. All tubes are packaged with caps attached and sterilized with irradiation.