Bag closures, WeLoc® Gripper / WeLoc® Gripper FD
Leverandør: WELOC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S1410001 S1410004 S1410024 S1410002
312-0112EA 2460 DKK
312-0112 312-0113 312-0114 312-0119
Bag closures, WeLoc® Gripper / WeLoc® Gripper FD
Klemmer Universalklemmer
The WeLoc® Gripper is the most powerful WeLoc® closure. It is manufactured from DuPont® Polyamide, reinforced with glass fibre which further enhances its strength and durability. The Gripper is designed to cope with the toughest environments and is therefore excellent for many industrial applications, where a stronger seal is often required.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Manufactured in Sweden under strict hygiene conditions
  • Clips can be sterilised via autoclave, EtO or gamma irradiation before being re-used
The WeLoc® Gripper FD is a powerful sack closure made from DuPont® Polyamide 6.6. It is fully metal and X-ray detectable and therefore ideal for use in the food and pharmaceutical industry.