Clips for 4" chip trays, H 44 Series
Leverandør: ENTEGRIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H44-04-0615 H44-041-61C02 H44-04B-61C02 H44-041-0615
216-1664EA 0 DKK
216-1664 216-1666 216-1671 216-1665
Clips for 4" chip trays, H 44 Series
Klemmer Universalklemmer
Interlocking tabs press on the center of the tray for secure contact between the tray and cover.
Advarsel: Articles may not be available in all countries. Please contact your local Avantor office.